missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol 95% (v/v), Extra Pure, SLR, Fisher Chemical™
41.46 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.
Use promo code "21953" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Plastic Container
Plastic-coated Glass Bottle
Specifications
Melting Point | -114°C |
Color | Colorless |
Boiling Point | 78°C |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Molecular Weight (g/mol) | 46.069 |
Formula Weight | 46.07g/mol |
Safety and Handling
Product Identifier
- Ethanol (95% v/v)
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'