missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 99.5%, Extra Dry, absolute, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10483195

Ethanol, 99.5%, Extra Dry, absolute, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10483195
1 L, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 10483195

Brand: Thermo Scientific Acros 397690010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Ethanol, >-99.5%, C2H6O, CAS Number-64-17-5, 7732-18-5, algrain, alcohol, ethyl hydroxide, ethyl hydrate, alcohol, ethyl alcohol, anhydrol, methylcarbinol, tecsol, grain alcohol, 100mL, CHEBI:16236, Colorless, 0.7900g/mL, 46.07, Extra Dry, Danger, GHS P Statement Highly flammable liquid and vapor.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Density 0.7900g/mL
Quantity 1 L
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Assay Percent Range 99.5%
Residue after Evaporation 0.001% max.
Linear Formula CH3CH2OH
UN Number 1170
Water 0.005% max.
Merck Index 15, 3814
Formula Weight 46.07
Percent Purity ≥99.5%
Grade Extra Dry
Chemical Name or Material Ethanol, Extra Dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1993606 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number397690010,397691000,397690025
Lot Number1904542 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number397690010,397691000,397690025
Lot Number1987584 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number397690010,397691000,397690025
Lot Number1904723 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number397690010,397691000,397690025
Lot Number1926554 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number397690010,397691000,397690025
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 99.5%, Extra Dry, absolute, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.