missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 99.8%, for HPLC, absolute, Thermo Scientific Chemicals Product Code.: 12347163

Ethanol, 99.8%, for HPLC, absolute, Thermo Scientific Chemicals

Product Code. 12347163
2.5 L
Click to view available options
Quantity:
2.5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 12347163

Brand: Thermo Scientific Acros 445740025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-17-5, 7732-18-5
Quantity 2.5 L
Molecular Formula C2H6O
Linear Formula CH3CH2OH
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Formula Weight 46.07
Grade HPLC
Chemical Name or Material Ethanol, absolute
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453124 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number445740010,445740025
Lot NumberA0451771 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number445740010,445740025
Lot NumberA0444268 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number
Lot NumberA0443094 Certificate TypeCertificate of Analysis Date09/08/2022 Catalog Number
Lot NumberA0437962 Certificate TypeCertificate of Analysis Date16/03/2022 Catalog Number
compliance-icons
Product Identifier
  • (±)-2-(2-Pyrrolidinyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 24126

    Expires2025-02-13

    Days

    22

    Hours

    4

    Minutes

    11

    Seconds

    5

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 99.8%, for HPLC, absolute, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.