missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol 99%+, Absolute, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Bottle
Plastic Container
Description
Ethanol 99%+, Absolute, Extra Pure, SLR
Specifications
Specifications
Melting Point | -114°C |
Color | Colorless |
Boiling Point | 78°C |
Quantity | 25 L |
Packaging | Plastic Container |
Physical Form | Liquid |
Vapor Pressure | 59 hPa at 20°C |
Viscosity | 1.2 mPaS at 20°C |
Molecular Weight (g/mol) | 46.069 |
Formula Weight | 46.07g/mol |
Safety and Handling
Product Identifier
- Ethanol 99+%, absolute
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction