missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol absolute, BAKER ANALYZED™ ACS, J.T. Baker™
Click to view available options
Quantity:
250 mL
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
Specifications
Specifications
Melting Point | -117°C |
Color | Max. 10 (APHA) |
Density | 0.7895 g/cm³ (20°C) |
Boiling Point | 78.3°C (1013 hPa) |
Quantity | 250 mL |
Flash Point | 12°C |
Packaging | Glass Bottle |
Assay Percent Range | min. 99.9% (v/v) |
Residue after Evaporation | max. 0.001% |
Titratable Acid | max. 0.0005 meq/g |
Show More |
Safety and Handling

Product Identifier
- Ethanol Absolute
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction