missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, Absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC), Honeywell™
Chemical Identifiers
CAS | 64-17-5, 78-93-3 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Chemical Identifiers
64-17-5, 78-93-3 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
-114°C (literature) | |
≥90.0000%, ≥1.0000% | |
C2H6O | |
MFCD00003568 | |
1718733 | |
Completely souble in water | |
CCO | |
46.069 | |
CHEBI:16236 | |
>98% | |
78°C to 79°C | |
12°C | |
Poly Bottle | |
Ethanol, Absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC) |
64-17-5, 78-93-3 | |
≤100.0000%, <5.0000% | |
CH3CH2OH | |
UN1170 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46 | |
0.789 g/mL at 25 °C(lit.) | |
19% (60°F) | |
683°F | |
44.6mmHg (20°C) |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319
Danger
missing translation for 'shelfLife' : 1800 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethanol
Recommended Storage : Room Temp