missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ethanol (EtOH), 20%

Gibco 20% Ethanol (EtOH) is sterile-filtered, packaged in bioprocess containers, and stocked to be available when you need it. Sourcing 20% EtOH can streamline your workload for equipment sanitization and column storage, allowing you to focus on the key steps in your downstream processes.

Brand:  Gibco™ A5803203

 View more versions of this product

Product Code. 18004090

  • £5712.37 / 200 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ethanol (EtOH) and bulk Water for Injection (WFI) are mixed to produce solutions that meet the most stringent quality control expectations in accordance with the pharmaceutical, biopharma, and diagnostic industries. Gibco 20% Ethanol is manufactured to cGMP standards at an ISO certified facility, promoting quality and convenience for every stage of a biologic’s pipeline.

Product features:
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Product specifications are well-defined, with the established solutions ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Meets workflow needs for all stages of biologic development and manufacturing
• Streamlines in-house processes related to formulation, hydration, testing, and validation
• Mitigates space required for buffer preparation and storage
• Eases the time requirements dedicated for in-house buffer preparation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ethyl Alcohol
200 L
Liquid
20%
20% EtOH
Clear
Drum
GMP - 21 CFR 820, ISO 13485
0.2 μm Sterile-filtered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.