missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, puriss., 96.0 to 97.2%, Honeywell™

£608.82 - £1017.87

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15662480
View Documents
Honeywell Riedel-de Haën
24105-4X2.5L
4 x 2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15672480
View Documents
Honeywell Riedel-de Haën
24105-4X5L
4 x 5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

64-17-5
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
C2H6O
MFCD00003568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO

Specifications

64-17-5
19% (60°F)
683°F
CH3CH2OH
44.6mmHg (20°C)
1718733
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.07g/mol
puriss.
0.805 to 0.812g/mL at 20°C, 0.789g/mL at 25°C
14°C (57.2°F)
C2H6O
MFCD00003568
UN1170
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
96.0 to 97.2%
Ethanol, Meets analytical specification of Ph.Eur.BP
Videos
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

H225-H319

missing translation for 'dotInformation' : class 3 - PG 2 - Ethanol

missing translation for 'einecsNumber' : 200-578-6

missing translation for 'rtecsNumber' : KQ6300000

SDS
Documents

Documents

Product Certifications