missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss., meets analytical spec. of BP, Ph. Eur., 96% (v/v), for license holders only, Honeywell Riedel-de Haën™
BIOPHARMA

£86.36 - £1518.49

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 8
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15622490
View Documents
Honeywell-Riedel-de Haen
24106-1L-GL
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15612490
View Documents
Honeywell-Riedel-de Haen
24106-1L
1 L Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15632490
BIOPHARMA
View Documents Promotion Details
Honeywell-Riedel-de Haen
24106-2.5L
2.5 L Plastic Bottle
£271.11
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15672490
BIOPHARMA
View Documents
Honeywell-Riedel-de Haen
24106-5L
5 L Poly Bottle
£492.91
5 L
Please to purchase this item. Need a web account? Register with us today!
15652490
BIOPHARMA
View Documents
Honeywell-Riedel-de Haen
24106-4X2.5L
4 x 2.5 L Plastic Bottle
£908.06
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15662490
BIOPHARMA
View Documents
Honeywell-Riedel-de Haen
24106-4X5L
4 x 5 L Poly Bottle
£1518.49
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15642490
View Documents
Honeywell-Riedel-de Haen
24106-30KG
30 kg Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15602490
View Documents
Honeywell-Riedel-de Haen
24106-160KG
160 kg Metal Drum This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

64-17-5
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

-114°C (literature)
100.0000%
78°C to 79°C
C2H6O
MFCD00003530
1718733
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46
puriss.
64-17-5
0.789 g/mL at 25 °C(lit.)
12°C
NaHSO3
UN1170
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
96%
Ethanol (Untaxed), Puriss., meets analytical specification of BP, Ph. Eur., 96% (v/v), for license holders only
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1800 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethanol

missing translation for 'einecsNumber' : 200-578-6

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications