missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss. p.a., ACS reagent, prima fine spirit, without additive, F15, ∼96%, for license holders only, Honeywell Riedel-de Haën™
47.92 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.
Use promo code "21953" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
4 x 2.5 L
6 x 1 L
Specifications
Density | 0.789g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 19% (60°F) |
Flash Point | 14°C (57.2°F) |
Ignition Point | 683°F |
Physical Form | Prima Fine Spirit |
Linear Formula | CH3CH2OH |
Vapor Pressure | 44.6mmHg (20°C) |
UN Number | UN1170 |
Beilstein | 1718733 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Ethanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ethanol; ethyl alcohol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'