missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss. p.a., ACS reagent, absolute alcohol, without additive, A15, ≥99.8%, for license holders only, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
25 mL
4 x 25 mL
1 L
2.5 L
25 L
4 x 2.5 L
6 x 1 L
Specifications
Specifications
Density | 0.789g/mL at 25°C, 0.790g/mL at 20°C |
Quantity | 25 mL |
Explosion Limit | 19% (60°F) |
Flash Point | 14°C (57.2°F) |
Ignition Point | 683°F |
Physical Form | Absolute Alcohol |
Linear Formula | CH3CH2OH |
MDL Number | MFCD00003568 |
Vapor Pressure | 44.6mmHg (20°C) |
UN Number | UN1170 |
Show More |
Safety and Handling
Product Identifier
- Ethanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'