missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, purum, ≥99.8%, Honeywell™
£34.94 - £558.90
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
15670640
|
Honeywell Research Chemicals
02883-1L |
1 L | |||||||
15680640
|
Honeywell Research Chemicals
02883-2.5L |
2.5 L |
£105.18
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15610650
|
Honeywell Research Chemicals
02883-6X1L |
6 x 1 L | |||||||
15600650
|
Honeywell Research Chemicals
02883-4X2.5L |
4 x 2.5 L |
£362.85
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15690640
|
Honeywell Research Chemicals
02883-25L |
25 L | |||||||
Description
Identifiants chimiques
64-17-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Spécification
64-17-5 | |
CH3CH2OH | |
UN1170 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
≥99.8% | |
0.789g/mL at 25°C | |
14°C (57.2°F) | |
Absolute Alcohol | |
Ethanol |
C2H6O | |
MFCD00003568 | |
1718733 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol | |
purum | |
19% (60°F) | |
683°F | |
44.6mmHg (20°C) |
Sécurité et traitement
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319
missing translation for 'dotInformation' : class 3 - PG 2 - Ethanol
missing translation for 'einecsNumber' : 200-578-6
missing translation for 'rtecsNumber' : KQ6300000
Identifiant produit
Mention d'avertissement
Catégorie de danger
Mention de danger
Conseils de prudence
- Ethanol
Mention d'avertissement
- Danger
Catégorie de danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Mention de danger
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.