missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Solution, Denatured, Sterile, Made with USP Water for Injection, 70%, BAKER™, J.T. Baker™
403.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 gL
Packaging:
Poly Bottle
Specifications
CAS | 64-17-5 |
Quantity | 1 gL |
Flash Point | 25.00°C |
Packaging | Poly Bottle |
Specific Gravity | 1L = 0.88kg |
Molecular Formula | C2H6O |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
IUPAC Name | ethanol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'