missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), CHROMASOLV™, absolute, for gradient elution, sales not in Germany, ≥99.9%, for license holders only, Honeywell Riedel-de Haen

Product Code. 15654730
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C (literature)
CAS Max % 100.0000%
Density 0.789 g/mL at 25 °C(lit.)
Boiling Point 78°C to 79°C
Quantity 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Formula Weight 46
Percent Purity >99.9%
Chemical Name or Material Ethanol (Untaxed), CHROMASOLV™, absolute, for gradient elution, sales not in Germany, ≥99.9%, for license holders only
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'