missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 96% (v/v), for license holders only, Honeywell Riedel-de Haën™
482.91 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.
Use promo code "21953" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
Stainless Steel Can
Specifications
Melting Point | -114°C (literature) |
CAS Max % | 100.0000% |
Linear Formula | CH2Cl2 |
UN Number | UN1170 |
Formula Weight | 46 |
Percent Purity | 96% |
Grade | ACS Reagent/Reagent Ph. Eur. |
Density | 0.789 g/mL at 25 °C(lit.) |
Boiling Point | 78°C to 79°C |
Quantity | 5 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'