missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanolamine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
Solubility
Miscible with water.
Notes
Store it away from heat, spark, open flames, hot surfaces and smoke. The storage area should be safe and enclosed.
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
Solubility
Miscible with water.
Notes
Store it away from heat, spark, open flames, hot surfaces and smoke. The storage area should be safe and enclosed.
Specifications
Melting Point | 10°C |
Density | 1.012 |
Boiling Point | 169°C to 170°C |
Flash Point | 93°C (199°F) |
Odor | Ammonia-like |
Linear Formula | HOCH2CH2NH2 |
Refractive Index | 1.454 |
Quantity | 2500 g |
UN Number | UN2491 |
Beilstein | 505944 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'