Learn More
Ethanolamine, 99%, H2O 0.5% max, Thermo Scientific Chemicals
£35.90 - £97.95
Chemical Identifiers
CAS | 141-43-5 |
---|---|
Molecular Formula | C2H7NO |
Molecular Weight (g/mol) | 61.08 |
MDL Number | MFCD00008183 |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
Synonym | ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
IUPAC Name | 2-aminoethanol |
SMILES | NCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368097
|
Thermo Scientific Alfa Aesar
022793.30 |
250 g |
£35.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358097
|
Thermo Scientific Alfa Aesar
022793.A1 |
1 kg | |||||||
Description
Ethanolamine is used as a surfactant, fluorimetric reagent, and to remove CO2 and H2S from natural gas and other gases, functional fluids, intermediates. It is also used as feedstock in the production of detergents, emulsifiers, polishes, pharmaceuticals, corrosion inhibitors, chemical intermediates, ion exchange agents, plating agents and surface treating agents, electrical and electronic products, paper products, personal care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-43-5 | |
61.08 | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
700 | |
2-aminoethanol |
C2H7NO | |
MFCD00008183 | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
CHEBI:16000 | |
NCCO |
Specifications
141-43-5 | |
1.012 g/mL | |
93°C (199°F) | |
C2H7NO | |
MFCD00008183 | |
505944 | |
14,3727 | |
Soluble in benzene,ether,carbon tetrachloride. | |
NCCO | |
61.08 | |
CHEBI:16000 | |
99% | |
Liquid |
10°C | |
169°C to 170°C | |
Ammonia-like | |
1.454 | |
UN2491 | |
Air sensitive; Hygroscopic | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
2-aminoethanol | |
700 | |
61.08 | |
H2O 0.5% max. | |
Ethanolamine |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE
missing translation for 'einecsNumber' : 205-483-3
missing translation for 'rtecsNumber' : KJ5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only