missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine hydrochloride, 98+%, Thermo Scientific Chemicals

£27.84 - £124.62

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.542
MDL Number MFCD00012892
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
Synonym 2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
PubChem CID 74819
IUPAC Name 2-aminoethanol;hydrochloride
SMILES C(CO)N.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313466
View Documents
Thermo Scientific Alfa Aesar
L14322.22
100 g
£27.84
100g
Please to purchase this item. Need a web account? Register with us today!
11323466
View Documents
Thermo Scientific Alfa Aesar
L14322.36
500 g
£124.62
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2002-24-6
97.542
PMUNIMVZCACZBB-UHFFFAOYSA-N
74819
C(CO)N.Cl
C2H8ClNO
MFCD00012892
2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
2-aminoethanol;hydrochloride

Specifications

2002-24-6
C2H8ClNO
3542892
14,3727
PMUNIMVZCACZBB-UHFFFAOYSA-N
2-aminoethanol;hydrochloride
74819
≥98%
83°C to 86°C
MFCD00012892
Hygroscopic
2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
C(CO)N.Cl
97.542
97.55
Ethanolamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 217-900-6

missing translation for 'rtecsNumber' : KJ6370000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only