missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ether, Ultra Low Water, Contains No Preservative, BakerDRY™, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
Specifications
Specifications
Flash Point | −45.00°C |
Packaging | Narrow Mouth Amber Glass Bottle, w/ Septum |
Quantity | 1 L |
Specific Gravity | 1L = 0.71kg |
Formula Weight | 74.12 |
Grade | BakerDRY™ |
Chemical Name or Material | Ether, Ultra Low Water, Contains No Preservative |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'