missing translation for 'onlineSavingsMsg'
Learn More

Ether, ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 15518564 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15518564

Brand: Avantor J.T.Baker 925902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use

Specifications

CAS 60-29-7
Flash Point −45°C(−113°F)
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C4H10O
MDL Number MFCD00011646
Quantity 1 L
Specific Gravity 1L = 0.71kg
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12
Percent Purity ≥99.0%
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.