Learn More
Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
Brand: Fisher Bioreagents BP1302-10
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 32
Additional Details : CAS Number : 1239-45-8 Weight : 0.01000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III
Description
Amber Poly Bottle; 10mL
Ethidium Bromide is an intercalating dye used in the visualization of DNA and RNA in electrophoresis gels and cesium chloride density gradients.Specifications
Ethidium Bromide | |
1% Solution | |
DNase-, RNase- and Protease-Free | |
MFCD00011724 | |
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l | |
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12 | |
394.32 | |
CHEBI:4883 | |
Molecular Biology | |
10 mL |
Red | |
1239-45-8,7732-18-5 | |
C21H20BrN3 | |
15, 4769 | |
ZMMJGEGLRURXTF-UHFFFAOYSA-N | |
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide | |
14710 | |
394.33 | |
Clear Bottle | |
Liquid |
Safety and Handling
- Ethidium bromide, 1% Solution/Molecular Biology
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
Hazard Statement
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 3, 8-Diamino-1-ethyl-6-phenylphenantridinium bromide