missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)

Product Code. 15430527
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Synonym 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide

Specifications

Color Red to purple
Formula Weight 394.32
Beilstein 3642536
Melting Point ca. 261°
Odor Weak
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Product Type Ethidium Bromide, Electrophoresis Reagent
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular Weight (g/mol) 394.32
Purity dry weight
Quantity 5 g
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.