missing translation for 'onlineSavingsMsg'
Learn More

Ethisterone, 98%, Thermo Scientific™

Product Code. 15445779
Click to view available options
Quantity:
5g
25g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
soluble in chloroform, DMSO (0.05 mg/ml), acetone (slightly ), ethanol (<1mg/mL at 25°C), and water (<1mg/mL at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-03-7
Molecular Formula C21H28O2
Molecular Weight (g/mol) 312.453
MDL Number MFCD00003656
InChI Key CHNXZKVNWQUJIB-SJFWLOONSA-N
Synonym pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one
PubChem CID 12016247
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C

Specifications

Quantity 25g
Beilstein 1889895
Merck Index 14,3741
Solubility Information soluble in chloroform,DMSO (0.05mg/ml),acetone (slightly ),ethanol (<1mg/ml at 25°C),and water (<1mg/ml at 25°C).
Formula Weight 312.45
Percent Purity 98%
Chemical Name or Material Ethisterone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.