missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Boc-3-benzylpiperidine-3-carboxylate, 95%, Thermo Scientific™

Product Code. 15486847
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170842-80-5
Molecular Formula C20H29NO4
Molecular Weight (g/mol) 347.46
MDL Number MFCD11500859
InChI Key ZTGGNYIHGLOCJH-UHFFFAOYNA-N
Synonym 1-tert-butyl 3-ethyl 3-benzylpiperidine-1,3-dicarboxylate, ethyl 1-boc-3-benzylpiperidine-3-carboxylate, 3-benzylpiperidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, +/--n-t-butoxycarbonyl-3-carboethoxy-3-phenylmethyl-piperidine, 3-benzylpiperidine-1,3-dicarboxylic acid1-tert-butyl ester 3-ethyl ester, 1,3-piperidinedicarboxylic acid, 3-phenylmethyl-, 1-1,1-dimethylethyl 3-ethyl ester, 1,3-piperidinedicarboxylicacid,3-phenylmethyl-,1-1,1-dimethylethyl 3-ethylester
PubChem CID 19696502
IUPAC Name 1-O-tert-butyl 3-O-ethyl 3-benzylpiperidine-1,3-dicarboxylate
SMILES CCOC(=O)C1(CC2=CC=CC=C2)CCCN(C1)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Formula Weight 347.45
Percent Purity 95%
Chemical Name or Material Ethyl 1-Boc-3-benzylpiperidine-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'