missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals

Product Code. 10754862
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35120-18-4
Molecular Formula C7H11BrO2
Molecular Weight (g/mol) 207.06
MDL Number MFCD00001322
InChI Key UTVNSHXHFRIXMM-UHFFFAOYSA-N
Synonym ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane
PubChem CID 97595
IUPAC Name ethyl 1-bromocyclobutane-1-carboxylate
SMILES CCOC(=O)C1(CCC1)Br

Specifications

Color Colorless
Density 1.2790g/mL
Boiling Point 85°C to 88°C (12.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula BrC4H6CO2C2H5
Quantity 5 g
Specific Gravity 1.279
Formula Weight 207.06
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl 1-bromocyclobutanecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 1-bromocyclobutanecarboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'