Learn More
Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals
£31.80 - £109.00
Chemical Identifiers
CAS | 35120-18-4 |
---|---|
Molecular Formula | C7H11BrO2 |
Molecular Weight (g/mol) | 207.06 |
MDL Number | MFCD00001322 |
InChI Key | UTVNSHXHFRIXMM-UHFFFAOYSA-N |
Synonym | ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane |
PubChem CID | 97595 |
IUPAC Name | ethyl 1-bromocyclobutane-1-carboxylate |
SMILES | CCOC(=O)C1(CCC1)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10422770
|
Thermo Scientific Acros
188710010 |
1 g | Glass bottle |
£31.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10754862
|
Thermo Scientific Acros
188710050 |
5 g | Glass bottle |
£109.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35120-18-4 | |
207.06 | |
UTVNSHXHFRIXMM-UHFFFAOYSA-N | |
97595 | |
CCOC(=O)C1(CCC1)Br |
C7H11BrO2 | |
MFCD00001322 | |
ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane | |
ethyl 1-bromocyclobutane-1-carboxylate |
Specifications
35120-18-4 | |
1.2790g/mL | |
79°C | |
95% min. (GC) | |
BrC4H6CO2C2H5 | |
1.279 | |
UTVNSHXHFRIXMM-UHFFFAOYSA-N | |
ethyl 1-bromocyclobutane-1-carboxylate | |
97595 | |
96% | |
Ethyl 1-bromocyclobutanecarboxylate |
Colorless | |
85°C to 88°C (12.0 mmHg) | |
Authentic | |
C7H11BrO2 | |
MFCD00001322 | |
ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane | |
CCOC(=O)C1(CCC1)Br | |
207.06 | |
207.06 | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 252-384-6
- Ethyl 1-bromocyclobutanecarboxylate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only