missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15497856
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-82-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD12022629
InChI Key AUUICUZWYQYTNO-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclopentanecarboxylate, ethyl 1-hydroxymethyl cyclopentane-1-carboxylate, 1-hydroxymethyl-cyclopentanecarboxylic acid ethyl ester
PubChem CID 46172884
IUPAC Name ethyl 1-(hydroxymethyl)cyclopentane-1-carboxylate
SMILES CCOC(=O)C1(CCCC1)CO

Specifications

Quantity 250 mg
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'