missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Description
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.
Notes
Incompatible with acids, bases and oxidizing agents.
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.
Notes
Incompatible with acids, bases and oxidizing agents.
Specifications
Specifications
Quantity | 250 mg |
Formula Weight | 144.16 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'