missing translation for 'onlineSavingsMsg'
Learn More
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate, Thermo Scientific™ Product Code.: 10066854

ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate, Thermo Scientific™

Product Code. 10066854
25 g
Click to view available options
Quantity:
25 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 10066854

Brand: Thermo Scientific Maybridge SB00646EB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 53266-94-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00005330
InChI Key SHQNGLYXRFCPGZ-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
PubChem CID 104454
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(N)=N1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Ethyl 2-(2-amino-1, 3-thiazol-4-yl)acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.