missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(4-pyridyl)thiazole-4-acetate, 97%, Thermo Scientific™

Product Code. 15430548
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80653-68-5
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD06739821
InChI Key JZBJUBBYJUUKQK-UHFFFAOYSA-N
Synonym ethyl 2-2-pyridin-4-yl thiazol-4-yl acetate, ethyl 2-4-pyridyl thiazole-4-acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethyl ester, 2-4-pyridyl thiazole-4-acetic acid ethyl ester, ethyl 2-2-pyridin-4-yl-1,3-thiazol-4-yl acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethylester
PubChem CID 7176350
IUPAC Name ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2

Specifications

Quantity 5 g
Formula Weight 248.3
Percent Purity 97%
Chemical Name or Material Ethyl 2-(4-pyridyl)thiazole-4-acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'