missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 15488395
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193140-71-5
Molecular Formula C5H9ClF3NO2
Molecular Weight (g/mol) 207.58
MDL Number MFCD14525479
InChI Key MNFISCBCWBUTTQ-UHFFFAOYNA-N
Synonym ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, 2-amino-3,3,3-trifluoro-propionic acid ethyl ester hydrochloride, ethyl 3,3,3-trifluoroalaninate, ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, ethyl 3,3,3-trifluoroalaninate-hydrogen chloride 1/1
PubChem CID 45790935
SMILES Cl.CCOC(=O)C(N)C(F)(F)F

Specifications

Melting Point ∼149°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
IUPAC Name ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
Formula Weight 207.58
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.