missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-(5-methyl-2-furyl)thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15438805
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 378196-87-3
Molecular Formula C12H13NO3S
Molecular Weight (g/mol) 251.3
MDL Number MFCD02654898
InChI Key BGFHMNNBBDLFDU-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-5-methylfuran-2-yl thiophene-3-carboxylate, 2-amino-4-5-methyl-furan-2-yl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4-5-methyl-2-furyl thiophene-3-carboxylate, ethyl 2-amino-4-5-methyl 2-furyl thiophene-3-carboxylate, ethyl 2-azanyl-4-5-methylfuran-2-yl thiophene-3-carboxylate, 2-amino-4-5-methyl-2-furanyl-3-thiophenecarboxylic acid ethyl ester, 2-amino-4-5-methyl-2-furyl thiophene-3-carboxylic acid ethyl ester
PubChem CID 706051
IUPAC Name ethyl 2-amino-4-(5-methylfuran-2-yl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=C(O2)C)N

Specifications

Melting Point 91°C to 92°C
Quantity 250 mg
Formula Weight 251.31
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-(5-methyl-2-furyl)thiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'