missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%

Product Code. p-7032597
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15448785

Brand: Thermo Scientific Alfa Aesar H34116.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40106-13-6 | C12H17NO2S | 239.333 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol and ethanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40106-13-6
Molecular Formula C12H17NO2S
Molecular Weight (g/mol) 239.333
MDL Number MFCD00216932
InChI Key XTUHIGALMIGZST-UHFFFAOYSA-N
Synonym ethyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, ethyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, ethyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, ethyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid ethyl ester, ethyl 2-aminocyclohepta b thiophene-3-carboxylate, aronis24232, ethyl 2-aminocycloheptathiophene-3-carboxylate, 2-amino-3-ethoxy-carbonyl-4,5-pentamethylenethiophene
PubChem CID 292644
IUPAC Name ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCCCC2)N

Specifications

Melting Point 87°C to 88°C
Quantity 1 g
Solubility Information Soluble in methanol and ethanol.
Formula Weight 239.33
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.