missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific Chemicals Product Code.: 15434766

Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. 15434766
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15434766

Brand: Thermo Scientific Alfa Aesar H59076.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.

Solubility
Sparingly soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 238749-50-3
Molecular Formula C9H8BrNO2S
Molecular Weight (g/mol) 274.132
MDL Number MFCD15730638
InChI Key KXGSKTSSCQBDOA-UHFFFAOYSA-N
Synonym ethyl 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylate, ethyl 2-bromothieno<3,2-b>pyrrole-5-carboxylate, 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4h-thieno 3.2-b pyrrole-5-carboxylate, 2-bromothieno 3,2-b-pyrrole-5-carboxylic acid ethyl ester, 2-bromothieno 3,2-b pyrrole-5-carboxylic acid ethyl esterShow MoreShow Less
PubChem CID 13164349
IUPAC Name ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=C(S2)Br

Specifications

Quantity 1 g
Beilstein 4426004
Solubility Information Sparingly soluble in water.
Formula Weight 274.13
Percent Purity 95%
Chemical Name or Material Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU26H021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59076.03
Lot NumberL11Y029 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59076.03
Lot NumberT23H015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59076.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.