missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%, Thermo Scientific Chemicals

Product Code. 10366924
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14337-43-0
Molecular Formula C4H6ClNO3
Molecular Weight (g/mol) 151.55
MDL Number MFCD00010209
InChI Key UXOLDCOJRAMLTQ-UTCJRWHESA-N
Synonym ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate
PubChem CID 9576005
IUPAC Name ethyl (2Z)-2-chloro-2-hydroxyiminoacetate
SMILES CCOC(=O)C(\Cl)=N\O

Specifications

Melting Point 70°C to 76°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 151.55
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-2-(hydroxyimino)acetate
compliance-icons compliance-icons
Product Identifier
  • Ethyl 2-chloro-2-(hydroxyimino)acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.