missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-isocyanato-4-methylvalerate, 97%, Thermo Scientific™

Product Code. 11342527
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64505-10-8
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
MDL Number MFCD00078351
InChI Key SSTQICMSRVMLQN-UHFFFAOYNA-N
Synonym ethyl 2-isocyanato-4-methylvalerate, ethyl 2-isocyanato-4-methyl valerate, ethyl 2-isocyanato-4-methyl-valerate, acmc-20al11, 2-isocyanato-4-methylpentanoic acid ethyl ester, pentanoic acid,2-isocyanato-4-methyl-, ethyl ester, 2s
PubChem CID 522132
IUPAC Name ethyl 2-isocyanato-4-methylpentanoate
SMILES CCOC(=O)C(CC(C)C)N=C=O

Specifications

Boiling Point 92°C to 93°C (8 mmHg)
Refractive Index 1.43
Quantity 5 g
UN Number UN2206
Beilstein 1240785
Sensitivity Moisture sensitive
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material Ethyl 2-isocyanato-4-methylvalerate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'