missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-octynoate, 98%, Thermo Scientific Chemicals

Product Code. 11498647
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10519-20-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00015286
InChI Key QPMDWYXUSMRVKT-UHFFFAOYSA-N
Synonym ethyl 2-octynoate, 2-octynoic acid, ethyl ester, 2-octynoic acid ethyl ester, ethyl 2-octynate, ethyl heptynecarbonate, unii-xth780u0n3, ethyl heptine carbonate, ethyl n-oct-2-ynoate, ethyl heptyne carboxylate, 2-octynoic acid, ethylester
PubChem CID 66338
IUPAC Name ethyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OCC

Specifications

Density 0.91
Boiling Point 85°C (6 mmHg)
Flash Point 99°C (210°F)
Linear Formula CH3(CH2)4C≡CCO2CH2CH3
Refractive Index 1.447
Quantity 5 g
Beilstein 1759822
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl 2-octynoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'