missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-oxocyclopentylacetate, 95%, Thermo Scientific Chemicals
£62.80 - £260.00
Chemical Identifiers
CAS | 20826-94-2 |
---|---|
Molecular Formula | C9H14O3 |
Molecular Weight (g/mol) | 170.21 |
MDL Number | MFCD00044715 |
InChI Key | PJMKFKUFBDXYEC-SSDOTTSWSA-N |
Synonym | ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate |
PubChem CID | 2723663 |
IUPAC Name | ethyl 2-(2-oxocyclopentyl)acetate |
SMILES | CCOC(=O)C[C@H]1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10275020
|
Thermo Scientific Acros
123040010 |
1 g | Glass bottle |
£62.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11985101
|
Thermo Scientific Acros
123040050 |
5 g | Glass bottle |
£260.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20826-94-2 | |
170.21 | |
PJMKFKUFBDXYEC-SSDOTTSWSA-N | |
2723663 | |
CCOC(=O)C[C@H]1CCCC1=O |
C9H14O3 | |
MFCD00044715 | |
ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate | |
ethyl 2-(2-oxocyclopentyl)acetate |
Specifications
20826-94-2 | |
Authentic | |
C9H14O3 | |
MFCD00044715 | |
PJMKFKUFBDXYEC-SSDOTTSWSA-N | |
ethyl 2-(2-oxocyclopentyl)acetate | |
2723663 | |
95% | |
Ethyl 2-oxocyclopentylacetate |
Colorless to Yellow | |
94% min. (GC) | |
1.4507 to 1.4527 | |
ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate | |
CCOC(=O)C[C@H]1CCCC1=O | |
170.21 | |
170.21 | |
Liquid |
RUO – Research Use Only