missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-dihydroxycinnamate, 97%, Thermo Scientific Chemicals

Product Code. 11496556
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66648-50-8
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00045754
InChI Key WDKYDMULARNCIS-GQCTYLIASA-N
Synonym ethyl caffeate, ethyl 3-3,4-dihydroxyphenyl acrylate, ethyl 3,4-dihydroxycinnamate, caffeic acid ethyl ester, ethyl trans-caffeate, unii-76gbb1ju5y, e-ethyl 3-3,4-dihydroxyphenyl acrylate, 3,4-dihydroxycinnamic acid ethyl ester, 76gbb1ju5y, chembl17347
PubChem CID 5317238
ChEBI CHEBI:69656
IUPAC Name ethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Quantity 25 g
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material Ethyl 3,4-dihydroxycinnamate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'