missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-dinitrobenzoate, 98%, Thermo Scientific™

Product Code. 11467184
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-71-3
Molecular Formula C9H8N2O6
Molecular Weight (g/mol) 240.17
MDL Number MFCD00007232
InChI Key IBQREHJPMPCXQA-UHFFFAOYSA-N
PubChem CID 69255
IUPAC Name ethyl 3,5-dinitrobenzoate
SMILES CCOC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 91°C to 92°C
Quantity 250 g
Beilstein 1995473
Synonym benzoic acid, 3,5-dinitro-, ethyl ester, acmc-20al01, ethyl 3,5-dinitrobenzoate, 3,5-dinitrobenzoic acid ethyl ester, 3,5-dinitro-benzoic acid ethyl ester, benzoic acid,5-dinitro-, ethyl ester, benzoic acid,3,5-dinitro-, ethyl ester
Formula Weight 240.17
Percent Purity 98%
Chemical Name or Material Ethyl 3,5-dinitrobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'