missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-amino-4,4,4-trifluorobutyrate, 95%, Thermo Scientific Chemicals Product Code.: 11323926

Ethyl 3-amino-4,4,4-trifluorobutyrate, 95%, Thermo Scientific Chemicals

Product Code. 11323926
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11323926

Brand: Thermo Scientific Alfa Aesar L17025.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170804-18-9
Molecular Formula C6H10F3NO2
Molecular Weight (g/mol) 185.146
MDL Number MFCD01075754
InChI Key KGUOZZWGWBBRRF-UHFFFAOYSA-N
Synonym ethyl 3-amino-4,4,4-trifluorobutyrate, ethyl-3-amino-4,4,4-trifluorobutyrate, acmc-20akzx, ethyl3-amino-4,4,4-trifluorobutyrate, 3-amino-4,4,4-trifluorobutyric acid ethyl esterShow MoreShow Less
PubChem CID 2774311
IUPAC Name ethyl 3-amino-4,4,4-trifluorobutanoate
SMILES CCOC(=O)CC(C(F)(F)F)N

Specifications

Boiling Point 70°C to 71°C (4mmHg)
Refractive Index 1.3831
UN Number UN2735
Beilstein 6136102
Quantity 250 mg
Formula Weight 185.15
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorobutyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-amino-4,4,4-trifluorobutyrate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.