missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%

Product Code. 10132960 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10132960

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 886-86-2 | C9H11NO2·CH4O3S | 261.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-86-2
Molecular Formula C9H11NO2·CH4O3S
Molecular Weight (g/mol) 261.29
MDL Number MFCD00013176
InChI Key FQZJYWMRQDKBQN-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt
PubChem CID 261501
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
SMILES CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O

Specifications

Melting Point 146.0°C to 150.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula H2NC6H4CO2C2H5·CH3SO3H
Quantity 50 g
Beilstein 14, 389
Merck Index 15, 9786
Solubility Information (5% in Water) Clear solution
Formula Weight 261.29
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Ethyl 3-aminobenzoate, methanesulfonic acid salt
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-aminobenzoate, methanesulfonic acid salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.