missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15439025
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 861208-16-4
Molecular Formula C10H8BrClN2O2
Molecular Weight (g/mol) 303.54
MDL Number MFCD06496148
InChI Key PEINUXVIHDZXNP-UHFFFAOYSA-N
Synonym ethyl 3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylate, ethyl 3-bromo-6-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, 3-bromo-6-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester hydrobromide
PubChem CID 5260614
IUPAC Name ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=C(Br)N2C=C(Cl)C=CC2=N1

Specifications

Melting Point 139°C to 141°C
Quantity 1 g
Formula Weight 303.54
Percent Purity 95%
Chemical Name or Material Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'