missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate, 95%, Thermo Scientific™

Product Code. 15448975
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215504-30-5
Molecular Formula C10H9BrN2O2
Molecular Weight (g/mol) 269.10
MDL Number MFCD12827886
InChI Key USZADKSDVGABGB-UHFFFAOYSA-N
PubChem CID 51063907
IUPAC Name ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
SMILES CCOC(=O)C1=CN2C(Br)=CN=C2C=C1

Specifications

Melting Point 95°C to 96°C
Quantity 250 mg
Synonym ethyl 3-bromoimidazo 1,2-a pyridine-6-carboxylate, ethyl3-bromoimidazo 1,2-a pyridine-6-carboxylate, ethyl 3-bromo-4-hydroimidazo 1,2-a pyridine-6-carboxylate, 3-bromo-imidazo 1,2-a pyridine-6-carboxylic acid ethyl ester, 3-bromoimidazo 1,2-a pyridine-6-carboxylic acid ethyl ester
Formula Weight 269.1
Percent Purity 95%
Chemical Name or Material Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'