Learn More
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
£37.20 - £344.00
Chemical Identifiers
CAS | 539-74-2 |
---|---|
Molecular Formula | C5H9BrO2 |
Molecular Weight (g/mol) | 181.03 |
MDL Number | MFCD00000251 |
InChI Key | FQTIYMRSUOADDK-UHFFFAOYSA-N |
Synonym | ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate |
PubChem CID | 68320 |
IUPAC Name | ethyl 3-bromopropanoate |
SMILES | CCOC(=O)CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499563
|
Thermo Scientific Alfa Aesar
A12402.14 |
25 g |
£37.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409573
|
Thermo Scientific Alfa Aesar
A12402.22 |
100 g |
£104.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419573
|
Thermo Scientific Alfa Aesar
A12402.36 |
500 g |
£344.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethyl 3-bromopropionate was used in the preparation of (3?,4?-dimethoxyphenyl)-5-oxopentanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.
Solubility
Soluble in water, alcohol, chloroform, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
Chemical Identifiers
539-74-2 | |
181.03 | |
FQTIYMRSUOADDK-UHFFFAOYSA-N | |
68320 | |
CCOC(=O)CCBr |
C5H9BrO2 | |
MFCD00000251 | |
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate | |
ethyl 3-bromopropanoate |
Specifications
539-74-2 | |
67°C to 71°C (10 mmHg) | |
C5H9BrO2 | |
MFCD00000251 | |
Light sensitive | |
Soluble in water,alcohol,chloroform,benzene. | |
CCOC(=O)CCBr | |
181.03 | |
181.03 | |
Ethyl 3-bromopropionate |
1.416 | |
79°C (174°F) | |
1.452 | |
1700933 | |
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate | |
FQTIYMRSUOADDK-UHFFFAOYSA-N | |
ethyl 3-bromopropanoate | |
68320 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 208-724-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Ethyl 3-bromopropionate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Ethyl 3-bromopropionate
RUO – Research Use Only