missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-chloro-2,2-dimethylpropanoate, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 3-chloro-2,2-dimethyl-propionic acid ethyl ester, propanoic acid,3-chloro-2,2-dimethyl-, ethyl ester, acmc-1c8oo, ethyl 3-chloro-2,2-dimethyl-propanoate, propanoic acid, 3-chloro-2,2-dimethyl-, ethyl ester |
Formula Weight | 164.63 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 3-chloro-2, 2-dimethylpropanoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'