missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals

Product Code. 15498715
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128073-16-5
Molecular Formula C9H7ClF3NO2
Molecular Weight (g/mol) 253.605
MDL Number MFCD06656414
InChI Key COMQYNZHBCNPNW-UHFFFAOYSA-N
Synonym ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
PubChem CID 22013558
IUPAC Name ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl

Specifications

Boiling Point 64°C to 66°C (0.008 mmHg)
Quantity 5 g
Formula Weight 253.6
Percent Purity 96%
Chemical Name or Material Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'