missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(chlorosulfonyl)isonicotinate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 249.67 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 3-(chlorosulfonyl)isonicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'