missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(dimethylamino)acrylate, 99%

Product Code. 11425707
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11425707

Brand: Thermo Scientific Alfa Aesar B24291.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 924-99-2 | C7H13NO2 | 143.186 g/mol

Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water 90 g/L (25°C).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-99-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00144269
InChI Key MVUMJYQUKKUOHO-AATRIKPKSA-N
Synonym ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
PubChem CID 5369162
IUPAC Name ethyl (E)-3-(dimethylamino)prop-2-enoate
SMILES CCOC(=O)C=CN(C)C

Specifications

Melting Point 17°C to 18°C
Boiling Point 118°C to 121°C (10 mmHg)
Linear Formula (CH3)2NCH=CHCO2CH2CH3
Refractive Index 1.512
Quantity 50 g
Solubility Information Soluble in water 90g/L (25°C).
Formula Weight 143.19
Percent Purity 99%
Chemical Name or Material Ethyl 3-(dimethylamino)acrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.