missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-dimethylaminopropionate, 97%

Product Code. 11351317
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11351317

Brand: Thermo Scientific Alfa Aesar L01542.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20120-21-2 | C7H15NO2 | 145.20 g/mol

Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20120-21-2
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.20
MDL Number MFCD00015166
InChI Key IZMJWGJGVGRWBW-UHFFFAOYSA-N
Synonym ethyl 3-dimethylamino propanoate, ethyl 3-dimethylaminopropionate, ethyl 3-dimethylamino propionate, n,n-dimethyl-beta-alanine ethyl ester, ethyl 3-n,n-dimethylamino propionate, ethyl n,n-dimethyl-beta-alaninate, ethyl beta-dimethylamino propionate, beta-alanine, n,n-dimethyl-, ethyl ester, ethyl 3-dimethyl-aminopropionate, ethyl-3-n,n-dimethylamino propionate
PubChem CID 88372
IUPAC Name ethyl 3-(dimethylamino)propanoate
SMILES CCOC(=O)CCN(C)C

Specifications

Boiling Point 56°C to 57°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Amine-like
Quantity 5 g
UN Number UN1993
Beilstein 1702477
Formula Weight 145.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-dimethylaminopropionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.